accessibility menu, dialog, popup

UserName

CAS RN 13813-41-7

IIIHo

CAS RN 13813-41-7

IUPAC Name: holmium(3+) triiodide
Synonyms: holmium(3+) triiodide
Molecular Weight (g/mol): 545.64
Molecular Formula: HoI3
InChi Key: KXCRAPCRWWGWIW-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Ho+3]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results