CAS RN 1380672-07-0
CAS RN 1380672-07-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
G007-LK, MedChemExpress
MedChemExpress G007-LK is a potent and selective inhibitor of TNKS1 and TNKS2, with IC50s of 46 nM and 25 nM, respectively.