accessibility menu, dialog, popup

UserName

CAS RN 1373921-09-5

CH3BrFFFF

CAS RN 1373921-09-5

IUPAC Name: 5-(bromomethyl)-2-fluoro-1-methyl-3-(trifluoromethyl)benzene
Synonyms: 5-(bromomethyl)-2-fluoro-1-methyl-3-(trifluoromethyl)benzene
Molecular Weight (g/mol): 271.05
Molecular Formula: C9H7BrF4
InChi Key: YMILKGXWAUIFCT-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(CBr)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results