CAS RN 1373921-00-6
IUPAC Name:
1-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]methanamine
Synonyms:
1-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol):
207.17
Molecular Formula:
C9H9F4N
InChi Key:
URDNLWAWYZBLQA-UHFFFAOYSA-N
SMILES:
CC1=C(F)C(=CC(CN)=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-methyl-5-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
CAS: 1373921-00-6 Molecular Formula: C9H9F4N Molecular Weight (g/mol): 207.172 MDL Number: MFCD22201070 InChI Key: URDNLWAWYZBLQA-UHFFFAOYSA-N Synonym: 4-fluoro-3-methyl-5-trifluoromethyl benzylamine,1-4-fluoro-3-methyl-5-trifluoromethyl phenyl methanamine,4-fluoro-3-methyl-5-trifluoromethyl phenyl methanamine PubChem CID: 86277677 IUPAC Name: [4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]methanamine SMILES: CC1=C(C(=CC(=C1)CN)C(F)(F)F)F