CAS RN 1373920-82-1
CAS RN 1373920-82-1
IUPAC Name:
2-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]acetic acid
Synonyms:
[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol):
236.17
Molecular Formula:
C10H8F4O2
InChi Key:
OEPYHPGEWLYDHQ-UHFFFAOYSA-N
SMILES:
CC1=C(F)C(=CC(CC(O)=O)=C1)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-methyl-5-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
CAS: 1373920-82-1 Molecular Formula: C10H8F4O2 Molecular Weight (g/mol): 236.166 MDL Number: MFCD22201074 InChI Key: OEPYHPGEWLYDHQ-UHFFFAOYSA-N Synonym: 4-fluoro-3-methyl-5-trifluoromethyl phenylacetic acid,4-fluoro-3-methyl-5-trifluoromethyl phenyl acetic acid,2-4-fluoro-3-methyl-5-trifluoromethyl phenyl acetic acid PubChem CID: 86277681 IUPAC Name: 2-[4-fluoro-3-methyl-5-(trifluoromethyl)phenyl]acetic acid SMILES: CC1=C(C(=CC(=C1)CC(=O)O)C(F)(F)F)F