accessibility menu, dialog, popup

UserName

CH3FFFFNH2

CAS RN 1373920-72-9

IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)aniline
Synonyms: 4-fluoro-3-methyl-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 193.15
Molecular Formula: C8H7F4N
InChi Key: HNXLESNHAMSHBA-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC(=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results