CAS RN 1373920-63-8
IUPAC Name:
4-fluoro-3-methyl-5-(trifluoromethyl)benzoic acid
Synonyms:
4-fluoro-3-methyl-5-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
222.14
Molecular Formula:
C9H6F4O2
InChi Key:
JBDVVWPLFZFUTO-UHFFFAOYSA-N
SMILES:
CC1=C(F)C(=CC(=C1)C(O)=O)C(F)(F)F
1
–
1
of
1
results
1
4-Fluoro-3-methyl-5-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 1373920-63-8 Molecular Formula: C9H6F4O2 Molecular Weight (g/mol): 222.139 MDL Number: MFCD22201064 InChI Key: JBDVVWPLFZFUTO-UHFFFAOYSA-N Synonym: 4-fluoro-3-methyl-5-trifluoromethyl benzoic acid PubChem CID: 86277671 IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)benzoic acid SMILES: CC1=C(C(=CC(=C1)C(=O)O)C(F)(F)F)F