accessibility menu, dialog, popup

UserName

CH3OOHNOOHNH3C

CAS RN 13684-63-4

IUPAC Name: 3-[(methoxycarbonyl)amino]phenyl N-(3-methylphenyl)carbamate
Synonyms: phenmedipham
Molecular Weight (g/mol): 300.31
Molecular Formula: C16H16N2O4
InChi Key: IDOWTHOLJBTAFI-UHFFFAOYSA-N
SMILES: COC(=O)NC1=CC=CC(OC(=O)NC2=CC=CC(C)=C2)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results