accessibility menu, dialog, popup

UserName

CAS RN 136705-77-6

SOOOFFFPP(R)(R)(R)(R)CH3CH3H3CH3CRh(Z)(Z)

CAS RN 136705-77-6

IUPAC Name: 1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane cycloocta-1,5-diene rhodium trifluoromethanesulfonate
Synonyms: 1,5-cyclooctadiene 1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane rhodium triflate
Molecular Weight (g/mol): 722.63
Molecular Formula: C31H48F3O3P2RhS
InChi Key: XGPXBCKGQLCHDW-UHFFFAOYNA-M
SMILES: [Rh].[O-]S(=O)(=O)C(F)(F)F.C1CC=CCCC=C1.CCC1CCC(CC)P1C1=CC=CC=C1P1C(CC)CCC1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)