CAS RN 13612-34-5
CAS RN 13612-34-5
IUPAC Name:
2-(2,5-dimethylphenyl)acetate
Synonyms:
(2,5-dimethylphenyl)acetate
Molecular Weight (g/mol):
163.2
Molecular Formula:
C10H11O2
InChi Key:
RUSCTNYOPQOXDJ-UHFFFAOYSA-M
SMILES:
CC1=CC=C(C)C(CC([O-])=O)=C1
1
–
2
of
2
results
2
2,5-Dimethylphenylacetic acid, 98+%, Thermo Scientific™
CAS: 13612-34-5 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00014369 InChI Key: RUSCTNYOPQOXDJ-UHFFFAOYSA-N Synonym: 2,5-dimethylphenylacetic acid,2-2,5-dimethylphenyl acetic acid,2,5-dimethylphenyl acetic acid,2,5-xylylacetic acid,2,5-dimethylphenylaceticacid,2,5-dimethylbenzeneacetic acid,benzeneacetic acid, 2,5-dimethyl,pubchem22525,acmc-1bb4h PubChem CID: 83615 IUPAC Name: 2-(2,5-dimethylphenyl)acetic acid SMILES: CC1=CC(=C(C=C1)C)CC(=O)O