accessibility menu, dialog, popup

UserName

CAS RN 13569-60-3

H2OSmOClOOOOClOOOOClOOO

CAS RN 13569-60-3

IUPAC Name: samarium(3+) hydrate triperchlorate
Synonyms: samarium(3+) hydrate triperchlorate
Molecular Weight (g/mol): 466.71
Molecular Formula: Cl3H2O13Sm
InChi Key: ONSMNAINZIBBGR-UHFFFAOYSA-K
SMILES: O.[Sm+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results