accessibility menu, dialog, popup

UserName

CAS RN 13535-01-8

NH2NBr

CAS RN 13535-01-8

IUPAC Name: 5-bromopyridin-3-amine
Synonyms: 5-bromopyridin-3-amine
Molecular Weight (g/mol): 173.01
Molecular Formula: C5H5BrN2
InChi Key: MDQXGHBCDCOOSM-UHFFFAOYSA-N
SMILES: NC1=CN=CC(Br)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)