CAS RN 135326-22-6
CAS RN 135326-22-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Gadoxetate Disodium, MedChemExpress
MedChemExpress Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is a contrast agent in magnetic resonance imaging (MRI) of the hepatobiliary system, which accumulates in normal, functioning hepatocytes. Gadoxetate Disodium (Gd-EOB-DTPA Disodium) is used to evaluate focal liver lesions, such as hepatocellular carcinoma or liver metastasis on T1-weighted imaging.