CAS RN 1347814-89-4
CAS RN 1347814-89-4
IUPAC Name:
1-(cyanomethyl)-2-(pyridin-2-yl)-1H-imidazole-4,5-dicarbonitrile
Synonyms:
1-(cyanomethyl)-2-(pyridin-2-yl)imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol):
234.22
Molecular Formula:
C12H6N6
InChi Key:
CKIOVYFZWSZUDP-UHFFFAOYSA-N
SMILES:
N#CCN1C(=NC(C#N)=C1C#N)C1=CC=CC=N1
1
–
1
of
1
results
1
1-Cyanomethyl-4,5-dicyano-2-(2-pyridyl)imidazole, 97%, Thermo Scientific™
CAS: 1347814-89-4 Molecular Formula: C12H6N6 Molecular Weight (g/mol): 234.222 MDL Number: MFCD20265178 InChI Key: CKIOVYFZWSZUDP-UHFFFAOYSA-N Synonym: 1-cyanomethyl-4,5-dicyano-2-2-pyridyl imidazole,1-cyanomethyl-2-pyridin-2-yl imidazole-4,5-dicarbonitrile PubChem CID: 73995634 IUPAC Name: 1-(cyanomethyl)-2-pyridin-2-ylimidazole-4,5-dicarbonitrile SMILES: C1=CC=NC(=C1)C2=NC(=C(N2CC#N)C#N)C#N