accessibility menu, dialog, popup

UserName

CAS RN 13465-58-2

OSOOOOSOOOOSOOOSmSm

CAS RN 13465-58-2

IUPAC Name: disamarium(3+) trisulfate
Synonyms: disamarium(3+) trisulfate
Molecular Weight (g/mol): 588.89
Molecular Formula: O12S3Sm2
InChi Key: LVSITDBROURTQX-UHFFFAOYSA-H
SMILES: [Sm+3].[Sm+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)