accessibility menu, dialog, popup

UserName

CAS RN 1323966-31-9

CH3OFH2NCl

CAS RN 1323966-31-9

IUPAC Name: 4-chloro-3-ethoxy-2-fluoroaniline
Synonyms: 4-chloro-3-ethoxy-2-fluoroaniline
Molecular Weight (g/mol): 189.61
Molecular Formula: C8H9ClFNO
InChi Key: FVVBUBBCVRLCSJ-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC(N)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results