accessibility menu, dialog, popup

UserName

CAS RN 1323966-25-1

CH3FFFFOOH

CAS RN 1323966-25-1

IUPAC Name: 3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[2-fluoro-5-methyl-4-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 250.19
Molecular Formula: C11H10F4O2
InChi Key: ZXGHBPAJUBOZLW-UHFFFAOYSA-N
SMILES: CC1=CC(CCC(O)=O)=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results