CAS RN 1323966-20-6
CAS RN 1323966-20-6
IUPAC Name:
2-fluoro-5-methyl-4-(trifluoromethyl)benzonitrile
Synonyms:
2-fluoro-5-methyl-4-(trifluoromethyl)benzonitrile
Molecular Weight (g/mol):
203.14
Molecular Formula:
C9H5F4N
InChi Key:
ITMYXYMYSSFKNP-UHFFFAOYSA-N
SMILES:
CC1=C(C=C(F)C(=C1)C#N)C(F)(F)F
1
–
1
of
1
results
1
2-Fluoro-5-methyl-4-(trifluoromethyl)benzonitrile, 97%, Thermo Scientific™
CAS: 1323966-20-6 Molecular Formula: C9H5F4N Molecular Weight (g/mol): 203.14 MDL Number: MFCD19687195 InChI Key: ITMYXYMYSSFKNP-UHFFFAOYSA-N Synonym: 2-fluoro-5-methyl-4-trifluoromethyl benzonitrile PubChem CID: 66523484 IUPAC Name: 2-fluoro-5-methyl-4-(trifluoromethyl)benzonitrile SMILES: CC1=C(C=C(C(=C1)C#N)F)C(F)(F)F