accessibility menu, dialog, popup

UserName

FOFFFOHO

CAS RN 1323966-19-3

IUPAC Name: 2-[4-fluoro-2-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [4-fluoro-2-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 238.14
Molecular Formula: C9H6F4O3
InChi Key: FXCYHVBZPAWHHJ-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(OC(F)(F)F)C=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results