accessibility menu, dialog, popup

UserName

CAS RN 1323955-63-0

CH3OFOOHCl

CAS RN 1323955-63-0

IUPAC Name: 2-(4-chloro-3-ethoxy-2-fluorophenyl)acetic acid
Synonyms: (4-chloro-3-ethoxy-2-fluorophenyl)acetic acid
Molecular Weight (g/mol): 232.64
Molecular Formula: C10H10ClFO3
InChi Key: JLMJTUGWXPDCKF-UHFFFAOYSA-N
SMILES: CCOC1=C(Cl)C=CC(CC(O)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results