accessibility menu, dialog, popup

UserName

CAS RN 13195-79-4

BrBrOBr

CAS RN 13195-79-4

IUPAC Name: 2,2-dibromo-1-(4-bromophenyl)ethan-1-one
Synonyms: acetophenone, 2,2,4'-tribromo-
Molecular Weight (g/mol): 356.84
Molecular Formula: C8H5Br3O
InChi Key: KFTUNOVZJWIKFX-UHFFFAOYSA-N
SMILES: BrC(Br)C(=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results