accessibility menu, dialog, popup

UserName

CAS RN 1315-04-4

SSbSSSbSS

CAS RN 1315-04-4

IUPAC Name: [(disulfanylidenestibanyl)sulfanyl]stibanedithione
Synonyms: antimony pentasulfide
Molecular Weight (g/mol): 403.82
Molecular Formula: S5Sb2
InChi Key: PPKVREKQVQREQD-UHFFFAOYSA-N
SMILES: S=[Sb](=S)S[Sb](=S)=S
Grade 
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results