accessibility menu, dialog, popup

UserName

CAS RN 1310372-82-7

H3COSiNCOOCH3OCH3

CAS RN 1310372-82-7

IUPAC Name: (10-isocyanatodecyl)trimethoxysilane
Synonyms: (10-isocyanatodecyl)trimethoxysilane
Molecular Weight (g/mol): 303.47
Molecular Formula: C14H29NO4Si
InChi Key: GUOFKJBNQHCAKJ-UHFFFAOYSA-N
SMILES: CO[Si](CCCCCCCCCCN=C=O)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results