accessibility menu, dialog, popup

UserName

CAS RN 1310-82-3

OHRb

CAS RN 1310-82-3

IUPAC Name: rubidium(1+) hydroxide
Synonyms: rubidium hydroxide RbOH
Molecular Weight (g/mol): 102.48
Molecular Formula: HORb
InChi Key: CPRMKOQKXYSDML-UHFFFAOYSA-M
SMILES: [OH-].[Rb+]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results