accessibility menu, dialog, popup

UserName

SNNOOH

CAS RN 1301738-68-0

IUPAC Name: 4-phenyl-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylic acid
Synonyms: 4-phenyl-2-(pyridin-2-yl)-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 282.32
Molecular Formula: C15H10N2O2S
InChi Key: AFWYDGNOZTZZDG-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=C(S1)C1=CC=CC=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results