CAS RN 1284220-49-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(Difluoromethyl)-1h-pyrazol-3-amine, TRC
CAS: 1284220-49-0 Molecular Formula: C4H5F2N3 Molecular Weight (g/mol): 133.1 SMILES: Nc1cc([nH]n1)C(F)F