CAS RN 1276105-89-5
CAS RN 1276105-89-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
CNX-1351, MedChemExpress
MedChemExpress CNX-1351 is a potent and isoform-selective targeted covalent PI3Kα inhibitor with IC50 of 6.8 nM.