CAS RN 126926-34-9
CAS RN 126926-34-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(Hydroxymethyl)Benzamide (~85%), TRC
CAS: 126926-34-9 Molecular Formula: C8H9NO2 Molecular Weight (g/mol): 151.16 Synonym: 3-(Hydroxymethyl)benzamide IUPAC Name: 3-(hydroxymethyl)benzamide SMILES: NC(C1=CC=CC(CO)=C1)=O