accessibility menu, dialog, popup

UserName

H3COOHOOFFF(E/Z)

CAS RN 1262018-46-1

IUPAC Name: 3-[4-methoxy-3-(trifluoromethoxy)phenyl]prop-2-enoic acid
Synonyms: 3-[4-methoxy-3-(trifluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 262.18
Molecular Formula: C11H9F3O4
InChi Key: DWBGYFCPZATQOQ-UHFFFAOYSA-N
SMILES: COC1=C(OC(F)(F)F)C=C(C=CC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results