CAS RN 126-58-9
CAS RN 126-58-9
IUPAC Name:
2-{[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl}-2-(hydroxymethyl)propane-1,3-diol
Synonyms:
dipentaerythritol
Molecular Weight (g/mol):
254.28
Molecular Formula:
C10H22O7
InChi Key:
TXBCBTDQIULDIA-UHFFFAOYSA-N
SMILES:
OCC(CO)(CO)COCC(CO)(CO)CO
No Refinements
1
–
0
of
0
results
1
Dipentaerythritol, 80+%, technical
CAS: 126-58-9 Molecular Formula: C10H22O7 Molecular Weight (g/mol): 254.28 MDL Number: MFCD00004691 InChI Key: TXBCBTDQIULDIA-UHFFFAOYSA-N Synonym: dipentaerythritol,dipentek,2,2'-oxybis methylene bis 2-hydroxymethyl propane-1,3-diol,bis pentaerythritol,1,3-propanediol, 2,2'-oxybis methylene bis 2-hydroxymethyl,2,2,2',2'-tetrakis hydroxymethyl-3,3'-oxydipropan-1-ol,1,3-propanediol, 2,2'-oxybis-methylene,2-3-hydroxy-2,2-bis hydroxymethyl propoxy methyl-2-hydroxymethyl propane-1,3-diol PubChem CID: 31352 IUPAC Name: 2-[[3-hydroxy-2,2-bis(hydroxymethyl)propoxy]methyl]-2-(hydroxymethyl)propane-1,3-diol SMILES: C(C(CO)(CO)COCC(CO)(CO)CO)O