accessibility menu, dialog, popup

UserName

H3COClOOHNH2F

CAS RN 1256482-68-4

IUPAC Name: 2-amino-3-(6-chloro-2-fluoro-3-methoxyphenyl)propanoic acid
Synonyms: 2-amino-3-(6-chloro-2-fluoro-3-methoxyphenyl)propanoic acid
Molecular Weight (g/mol): 247.65
Molecular Formula: C10H11ClFNO3
InChi Key: VWUCVGXVDGKDBN-UHFFFAOYNA-N
SMILES: COC1=C(F)C(CC(N)C(O)=O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results