CAS RN 1252594-99-2
CAS RN 1252594-99-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ETP-46321, MedChemExpress
MedChemExpress ETP-46321 is a potent and orally bioavailable PI3Kα and PI3Kδ inhibitor with Kiapps of 2.3 and 14.2 nM, respectively.