accessibility menu, dialog, popup

UserName

CAS RN 125117-97-7

NHNSNH2Cl

CAS RN 125117-97-7

IUPAC Name: (6-chloropyridin-3-yl)thiourea
Synonyms: 6-chloropyridin-3-ylthiourea
Molecular Weight (g/mol): 187.65
Molecular Formula: C6H6ClN3S
InChi Key: KQNRFXLAUHMUSH-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CN=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results