accessibility menu, dialog, popup

UserName

H3CONFFFBr

CAS RN 124432-63-9

IUPAC Name: 3-bromo-2-methoxy-5-(trifluoromethyl)pyridine
Synonyms: 3-bromo-2-methoxy-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 256.02
Molecular Formula: C7H5BrF3NO
InChi Key: YUDYUCKEWCVXTE-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C(C=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results