accessibility menu, dialog, popup

UserName

SFFFFFFOHO

CAS RN 1240257-93-5

IUPAC Name: 2-[2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)phenyl]acetic acid
Synonyms: [2-fluoro-4-(pentafluoro-λ⁶-sulfanyl)phenyl]acetic acid
Molecular Weight (g/mol): 280.18
Molecular Formula: C8H6F6O2S
InChi Key: ZTDHXJDCNBDLPP-UHFFFAOYSA-N
SMILES: OC(=O)CC1=C(F)C=C(C=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results