accessibility menu, dialog, popup

UserName

CAS RN 1240257-80-0

OFFFFN

CAS RN 1240257-80-0

IUPAC Name: 2-[2-fluoro-4-(trifluoromethoxy)phenyl]acetonitrile
Synonyms: 2-[2-fluoro-4-(trifluoromethoxy)phenyl]acetonitrile
Molecular Weight (g/mol): 219.14
Molecular Formula: C9H5F4NO
InChi Key: CABIXKMVRXTRSN-UHFFFAOYSA-N
SMILES: FC1=C(CC#N)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results