accessibility menu, dialog, popup

UserName

SFFFFFBrO

CAS RN 1240257-22-0

IUPAC Name: 3-bromo-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Synonyms: 3-bromo-5-(pentafluoro-λ⁶-sulfanyl)benzaldehyde
Molecular Weight (g/mol): 311.06
Molecular Formula: C7H4BrF5OS
InChi Key: IPQVQQNYNAJWRU-UHFFFAOYSA-N
SMILES: FS(F)(F)(F)(F)C1=CC(Br)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results