accessibility menu, dialog, popup

UserName

CAS RN 1240256-77-2

IUPAC Name: 2-(difluoromethoxy)-5-fluorobenzamide
Synonyms: 2-(difluoromethoxy)-5-fluorobenzamide
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: YVAGFUXIMXZUHA-UHFFFAOYSA-N
SMILES: NC(=O)C1=C(OC(F)F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results