accessibility menu, dialog, popup

UserName

CAS RN 123843-66-3

H3COFNF

CAS RN 123843-66-3

IUPAC Name: 2,6-difluoro-4-methoxybenzonitrile
Synonyms: 2,6-difluoro-4-methoxybenzonitrile
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H5F2NO
InChi Key: CMGFDVIQHBAJHT-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(C#N)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results