accessibility menu, dialog, popup

UserName

CAS RN 1234-35-1

H2NNH2HN(S)NHOOOO

CAS RN 1234-35-1

IUPAC Name: (2S)-5-{[amino(iminiumyl)methyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoate
Synonyms: (2S)-5-{[amino(iminio)methyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoate
Molecular Weight (g/mol): 308.34
Molecular Formula: C14H20N4O4
InChi Key: SJSSFUMSAFMFNM-NSHDSACASA-N
SMILES: NC(=[NH2+])NCCC[C@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)