accessibility menu, dialog, popup

UserName

CH3SOONSOOHNOHNNOCH3OCH3N

CAS RN 122931-48-0

IUPAC Name: 1-(4,6-dimethoxypyrimidin-2-yl)-3-{[3-(ethanesulfonyl)pyridin-2-yl]sulfonyl}urea
Synonyms: rimsulfuron
Molecular Weight (g/mol): 431.44
Molecular Formula: C14H17N5O7S2
InChi Key: MEFOUWRMVYJCQC-UHFFFAOYSA-N
SMILES: CCS(=O)(=O)C1=CC=CN=C1S(=O)(=O)NC(=O)NC1=NC(OC)=CC(OC)=N1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results