accessibility menu, dialog, popup

UserName

CH3ONOOHBr(E/Z)

CAS RN 1228963-18-5

IUPAC Name: 3-(5-bromo-2-methoxypyridin-3-yl)prop-2-enoic acid
Synonyms: 3-(5-bromo-2-methoxypyridin-3-yl)prop-2-enoic acid
Molecular Weight (g/mol): 258.07
Molecular Formula: C9H8BrNO3
InChi Key: AJOCJTKIVWCVMY-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1C=CC(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results