CAS RN 12209-98-2
CAS RN 12209-98-2
IUPAC Name:
disodium oxostannanebis(olate) trihydrate
Synonyms:
disodium oxostannanebis(olate) trihydrate
Molecular Weight (g/mol):
266.73
Molecular Formula:
H6Na2O6Sn
InChi Key:
SFXJSNATBHJIDS-UHFFFAOYSA-N
SMILES:
O.O.O.[Na+].[Na+].[O-][Sn]([O-])=O
1
–
1
of
1
results
1
Sodium tin(IV) oxide trihydrate, 96%
CAS: 12209-98-2 Molecular Formula: H6Na2O6Sn Molecular Weight (g/mol): 266.732 MDL Number: MFCD00150189 InChI Key: SFXJSNATBHJIDS-UHFFFAOYSA-N Synonym: sodium stannate trihydrate,unii-nj7c1v83kg,nj7c1v83kg,sodium stannate inci,sodium stannate vandf,disodium stannate, trihydrate,sodium stannate iv trihydrate mi,2na.so3n.3h2o,sodiumstannatetrihydrate,sodium tin iv oxide trihydrate PubChem CID: 21977542 IUPAC Name: disodium;dioxido(oxo)tin;trihydrate SMILES: O.O.O.[O-][Sn](=O)[O-].[Na+].[Na+]