CAS RN 1218816-71-7
CAS RN 1218816-71-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
SK1-IN-1, MedChemExpress
MedChemExpress SK1-IN-1 is a potent sphingosine kinase 1 (SPHK1) inhibitor with an IC50 of 58 nM.