accessibility menu, dialog, popup

UserName

BOOCH3CH3H3CCH3NCH3

CAS RN 1217891-71-8

IUPAC Name: 9-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 9-methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Molecular Weight (g/mol): 307.2
Molecular Formula: C19H22BNO2
InChi Key: AVCKYKVGQYBADD-UHFFFAOYSA-N
SMILES: CN1C2=C(C=CC=C2)C2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)