accessibility menu, dialog, popup

UserName

CAS RN 1217500-67-8

BClNHOOH

CAS RN 1217500-67-8

IUPAC Name: (3-chloro-2-cyanophenyl)boronic acid
Synonyms: 3-chloro-2-cyanophenylboronic acid
Molecular Weight (g/mol): 181.38
Molecular Formula: C7H5BClNO2
InChi Key: NVFNOKJQOOUGTR-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C#N)C(Cl)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results