accessibility menu, dialog, popup

UserName

CAS RN 1212-29-9

IUPAC Name: 1,3-dicyclohexylthiourea
Synonyms: thiourea, N,N'-dicyclohexyl-
Molecular Weight (g/mol): 240.41
Molecular Formula: C13H24N2S
InChi Key: KAJICSGLHKRDLN-UHFFFAOYSA-N
SMILES: S=C(NC1CCCCC1)NC1CCCCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results