CAS RN 1210344-57-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ertugliflozin, TRC
CAS: 1210344-57-2 Molecular Formula: C22H25ClO7 Molecular Weight (g/mol): 436.88 Synonym: 1,6-Anhydro-1-C-[4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl]-5-C-(hydroxymethyl)-beta-L-idopyranose,MK 8835,PF 04971729,PF 04971729-00,Steglatro SMILES: CCOc1ccc(Cc2cc(ccc2Cl)[C@]34OC[C@](CO)(O3)[C@@H](O)[C@H](O)[C@H]4O)cc1
2
Ertugliflozin, MedChemExpress
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.