accessibility menu, dialog, popup

UserName

CAS RN 121012-89-3

OOOOSrCF3CF3CF3CF3

CAS RN 121012-89-3

IUPAC Name: strontium(2+) bis(1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
Synonyms: strontium(2+) bis(1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 501.72
Molecular Formula: C10H2F12O4Sr
InChi Key: KEVXBFHXVKDJRZ-UHFFFAOYSA-L
SMILES: [Sr++].[O-]C(=CC(=O)C(F)(F)F)C(F)(F)F.[O-]C(=CC(=O)C(F)(F)F)C(F)(F)F
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results