accessibility menu, dialog, popup

UserName

CAS RN 1206593-28-3

CH3OOHOFFF

CAS RN 1206593-28-3

IUPAC Name: 2-[4-ethoxy-3-(trifluoromethyl)phenyl]acetic acid
Synonyms: [4-ethoxy-3-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 248.2
Molecular Formula: C11H11F3O3
InChi Key: WRQKZAJHRRGAJO-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(CC(O)=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results